missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FABP2 Protein

Product Code. p-7149681
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Unit Size:
1mg
25µg
5µg
This item is not returnable. View return policy

Product Code. 15918528

Brand: enQuireBio™ QP118345ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the FABP2 was constructed and used to recombinantly synthesize the protein.

Specifications

Name FABP2 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence AFDSTWKVDR SENYDKFMEK MGVNIVKRKL AAHDNLKLTI TQEGNKFTVK ESSAFRNIEV VFELGVTFNY NLADGTELRG TWSLEGNKLI GKFKRTDNGN ELNTVREIIG DELVQTYVYE GVEAKRIFKK D
Buffer FABP2 was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4.
Purity or Quality Grade Greater than 97.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.