missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FAM19A5 Protein

A cDNA sequence encoding the FAM19A5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11843-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958558

  • 131.00 € / 2µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

25817
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MKHHHHHHASQFLKEGQLAA GTCEIVTLDR DSSQPRRTIA RQTARCACRK GQIAGTTRAR PACVDARIIK TKQWCDMLPC LEGEGCDLLI NRSGWTCTQP GGRIKTTTVS
Greater than 95.0% as determined by SDS-PAGE.
FAM19A5 Protein
Research Use Only
FAM19A5
Human
His
FAM19A5 filtered (0.4microm) solution at a concentration of 0.2 mg/ml in 20mM Tris buffer, 50mM NaCl, pH 7.5 and 10% (w/v) glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers