missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FAM3C Protein

A cDNA sequence encoding the FAM3C was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11846-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928568

  • 203.00 € / 10µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FAM3C Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQVFEIKMDA SLGNLFARSA LDTAARSTKP PRYKCGISKA CPEKHFAFKM ASGAANVVGP KICLEDNVLM SGVKNNVGRG INVALANGKT GEVLDTKYFD MWGGDVAPFI EFLKAIQDGT IVLMGTYDDG ATKLNDEARR LIADLGSTSI TNLGFRDNWV FCGGKGIKTK SPFEQHIKNN KDTNKYEGWP EVVEMEGCIP QKQD
Greater than 90% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The FAM3C solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 200mM NaCl and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers