missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FETUB Protein

A cDNA sequence encoding the FETUB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11868-1mg


Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958638

  • 7540.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FETUB Protein
Research Use Only
Recombinant Protein
Human (HEK293)
HSHVCGAMSP PQLALNPSAL LSRGCNDSDV LAVAGFALRD INKDRKDGYV LRLNRVNDAQ EYRRGGLGSL FYLTLDVLET DCHVLRKKAW QDCGMRIFFE SVYGQCKAIF YMNNPSRVLY LAAYNCTLRP VSKKKIYMTC PDCPSSIPTD SSNHQVLEAA TESLAKYNNE NTSKQYSLFK VTRASSQWVV GPSYFVEYLI KESPCTKSQA SSCSLQSSDS VPVGLCKGSL TRTHWEKFVS VTCDFFESQA PATGSENSAV NQKPTNLPKV EESQQKNTPP TDSPSKAGPR GSVQYLPDLD DKNSQEKGPQ EAFPVHLDLT TNPQGETLDI SFLFLEPMEE KLVVLPFPKE KARTAECPGP AQNASPLVLP PHHHHHH
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FETUB filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers