missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FGF basic/FGF2/bFGF, Animal-Free Protein

Product Code. 18757136
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18757136 1 mg 1mg
18765456 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18757136 Supplier R&D Systems™ Supplier No. AFL371801M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Also available, GMP FGF basic/FGF2/bFGF Protein, Catalog # 3718-GMP! Animal-free and manufactured under GMP guidelines for therapeutic manufacturing applications.

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.1-0.6 ng/mL. The specific activity of Recombinant Human FGF basic/FGF2/bFGF is approximately 800 IU/μg, which is calibrated against human FGF basic/FGF2/bFGF WHO International Standard (NIBSC code: 90/712).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001997
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl.
Gene ID (Entrez) 2247
Molecular Weight (g/mol) M.W.-theoretical: 16 kDa
Quantity 1 mg
Source E. coli-derived human FGF basic/FGF2/bFGF protein Pro143-Ser288 Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein FGF basic/FGF2/bFGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.