missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Recombinant Human FGF basic Heat Stable Protein, CF

Product Code. p-200086905
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
50µg
This item is not returnable. View return policy

Product Code. 30067268

Brand: R&DSystems™ BTFGFBHS500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.0500-0.600 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001997.5
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HEPES and Sodium Sulfate with Trehalose.
Gene ID (Entrez) 2247
Molecular Weight (g/mol) M.W.-observed: 16-19 kDa, under reducing conditions., M.W.-theroretical: 17 kDa
Quantity 500 μg
Source E. coli-derived human FGF basic/FGF2/bFGF protein Ala135-Ser288 (with modifications)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Conjugate Unconjugated
Reconstitution Reconstitute 10 μg size at 100 μg/mL in sterile deionized water. Reconstitute all the other sizes at 500 μg/mL in sterile deionized water.
Protein FGF basic/FGF2/bFGF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.