missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FGF10 / KGF2 Protein

A DNA sequence encoding the Homo sapiens (Human) FGF10 / KGF2 Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6039-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959539

  • 505.00 € / 100µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004456.1
FGF10 / KGF2 Protein
Research Use Only
FGF10
Recombinant Protein
E. coli
CQALGQDMVS PEATNSSSSS FSSPSSAGRH VRSYNHLQGD VRWRKLFSFT KYFLKIEKNG KVSGTKKENC PYSILEITSV EIGVVAVKAI NSNYYLAMNK KGKLYGSKEF NNDCKLKERI EENGYNTYAS FNWQHNGRQM YVALNGKGAP RRGQKTRRKN TSAHFLPMVV HS
Greater than 80% as determined by SDS-PAGE.
2255
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers