missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF14 / SCA27 Protein
A DNA sequence encoding the Homo sapiens (Human) FGF14 / SCA27, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_004106.1 |
---|---|
Gene ID (Entrez) | 2259 |
Name | FGF14 / SCA27 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939559
|
enQuireBio™
QP6043-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959559
|
enQuireBio™
QP6043-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949559
|
enQuireBio™
QP6043-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969559
|
enQuireBio™
QP6043-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15979559
|
enQuireBio™
QP6043-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15989559
|
enQuireBio™
QP6043-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004106.1 | |
FGF14 / SCA27 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
2259 | |
Research Use Only | |
FGF14 | |
Recombinant Protein | |
E. coli | |
MVKPVPLFRR TDFKLLLCNH KDLFFLRVSK LLDCFSPKSM WFLWNIFSKG THMLQCLCGK SLKKNKNPTD PQLKGIVTRL YCRQGYYLQM HPDGALDGTK DDSTNSTLFN LIPVGLRVVA IQGVKTGLYI AMNGEGYLYP SELFTPECKF KESVFENYYV IYSSMLYRQQ ESGRAWFLGL NKEGQAMKGN RVKKTKPAAH FLPKPLEVAM YREPSLHDVG ETVPKPGVTP SKSTSASAIM NGGKPVNKSK TT | |
Greater than 80% as determined by SDS-PAGE. |