missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FGF21 / Fibroblast Growth Factor 21 Protein

Product Code. 15905819
Change view
Click to view available options
Quantity:
1 mg
20 μg
500 μg
Unit Size:
1mg
20µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15905819 20 μg 20µg
15915819 500 μg 500µg
15995809 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15905819 Supplier enQuireBio™ Supplier No. QP527220ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_061986.1
Gene ID (Entrez) 26291
Name FGF21 / Fibroblast Growth Factor 21 Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol FGF21
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MHPIPDSSPL . LQFGGQVRQ R. YLYTDDAQ QT. EAHLEIR EDG. TVGGAA DQSP. ESLLQ LKALK. PGVI QILGVK. TSR FLCQRPD. GA LYGSLHFD. P EACSFRELLL EDGYNVYQS.  EAHGLPLHLP . GNKSPHRDP A. PRGPARFL PL. PGLPPAL PEP. PGILAP QPPD. VGSSD PLSMV. GPSQ GRSPSY. AS
Buffer 10 mM sodium phosphate, 100 mM soldium chloride, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.