missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF22 Protein
Recombinant Protein
107.00 € - 6815.00 €
Specifications
Accession Number | NP_065688.1 |
---|---|
Gene ID (Entrez) | 27006 |
Name | FGF22 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15996009
|
enQuireBio™
QP5374-5UG |
5 μg |
107.00 €
5µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976009
|
enQuireBio™
QP5374-20UG |
20 μg |
252.00 €
20µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956009
|
enQuireBio™
QP5374-100UG |
100 μg |
920.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986009
|
enQuireBio™
QP5374-500UG |
500 μg |
4155.00 €
500µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966009
|
enQuireBio™
QP5374-1MG |
1 mg |
6815.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_065688.1 | |
FGF22 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MTPSASRGPR . SYPHLEGDV R. WRRLFSST HF. FLRVDPG GRV. QGTRWR HGQD. SILEI RSVHV. GVVV IKAVSS. GFY VAMNRRG. RL YGSRLYTVDC RFRERIEE. N GHNTYASQR. WRRRGQPMFL . ALDRRGGPR P. GGRTRRYH LS. AHFLPVL VS | |
Greater than 95% as determined by SDS-PAGE. |
27006 | |
Research Use Only | |
FGF22 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |