missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF8 Protein
Recombinant Protein
Brand: enQuireBio™ QP5301-25ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006110.1 | |
FGF8 Protein | |
25 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
At minimum, our recombinant Human FGF8 induces 3T3 cell proliferation at concentrations of less than 150 ng/ml (activity must be at least 6.7 x 10^3 units/mg). | |
Human | |
Untagged | |
5 mM sodium phosphate, 50mM soldium chloride, pH 7.5 |
2253 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
FGF8 | |
Recombinant Protein | |
E. coli | |
MQVTVQSSPN . FTQHVREQS L. VTDQLSRR LI. RTYQLYS RTS. GKHVQV LANK. RINAM AEDGD. PFAK LIVETD. TFG SRVRVRGAET GLYICMN. KK GKLIAKSN. G KGKDCVFTE. IVLENNYTAL . QNAKYEGWY M. AFTRKGRP RK. GSKTRQH QRE. VHFMKR LPRG. HHTTE QSLRFEFLNY PPFTR. SLRG SQRTWA. PEP R |