missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fibroleukin Protein

Product Code. 16029470
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16029470 100 μg 100µg
16039470 10 μg 10µg
16079470 50 μg 50µg
16059470 200 μg 200µg
16069470 500 μg 500µg
16049470 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16029470 Supplier enQuireBio™ Supplier No. QP7571ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Fibroleukin, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_006673.1
Gene ID (Entrez) 10875
Name Fibroleukin Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol FGL2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence NNETEEIKDE RAKDVCPVRL ESRGKCEEAG ECPYQVSLPP LTIQLPKQFS RIEEVFKEVQ NLKEIVNSLK KSCQDCKLQA DDNGDPGRNG LLLPSTGAPG EVGDNRVREL ESEVNKLSSE LKNAKEEINV LHGRLEKLNL VNMNNIENYV DSKVANLTFV VNSLDGKCSK CPSQEQIQSR PVQHLIYKDC SDYYAIGKRS SETYRVTPDP KNSSFEVYCD METMGGGWTV LQARLDGSTN FTRTWQDYKA GFGNLRREFW LGNDKIHLLT KSKEMILRID LEDFNGVELY ALYDQFYVAN EFLKYRLHVG NYNGTAGDAL RFNKHYNHDL KFFTTPDKDN DRYPSGNCGL YYSSGWWFDA CLSANLNGKY YHQKYRGVRN GIFWGTWPGV SEAHPGGYKS SFKEAKMMIR PKHFKP
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.