missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Flavokinase Protein
A cDNA sequence encoding the Flavokinase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11888-5ug
Additional Details : Weight : 0.01000kg
Specifications
Flavokinase Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPRADCIMRH LPYFCRGQVV RGFGRGSKQL GIPTANFPEQ VVDNLPADIS TGIYYGWASV GSGDVHKMVV SIGWNPYYKN TKKSMETHIM HTFKEDFYGE ILNVAIVGYL RPEKNFDSLE SLISAIQGDI EEAKKRLELP EHLKIKEDNF FQVSKSKIMNGH | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The Flavokinase solution containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |