missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Flt-3 Ligand/FLT3L Protein

Product Code. 18791388
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18791388 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18791388 Supplier R&D Systems™ Supplier No. 308FK01M/CF
  • ONLINE EXCLUSIVE PRICE
    6056.80 €
    5382.30 € / 1mg
    Save 674.50 €
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse Flt-3. The ED50 for this effect is 0.2-1 ng/mL.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAA17999.1
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA
Gene ID (Entrez) 2323
Molecular Weight (g/mol) 17.5 kDa (monomer)
Quantity 1 mg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Flt-3 Ligand/FLT3L protein Thr27-Pro185
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method
Gene Alias FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Biological Activity 0.2 to 1ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.