missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FSTL1 Protein

A cDNA sequence encoding the FSTL1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11905-HIS-HEK-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928758

  • 203.00 € / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

11167
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Human (HEK293)
MGSSHHHHHH SSGLVPRGSH MEEELRSKSK ICANVFCGAG RECAVTEKGE PTCLCIEQCK PHKRPVCGSN GKTYLNHCEL HRDACLTGSK IQVDYDGHCK EKKSVSPSAS PVVCYQSNRD ELRRRIIQWL EAEIIPDGWF SKGSNYSEIL DKYFKNFDNG DSRLDSSEFL KFVEQNETAI NITTYPDQEN NKLLRGLCVD ALIELSDENA DWKLSFQEFL KCLNPSFNPP EKKCALEDET YADGAETEVD CNRCVCACGN WVCTAMTCDG KNQKGAQTQT EEEMTRYVQE LQKHQETAEK TKRVSTKEI
Greater than 90% as determined by SDS-PAGE.
FSTL1 Protein
Research Use Only
FSTL1
Human
His
The FSTL1 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers