missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FTSJ2 Protein
A cDNA sequence encoding the FTSJ2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11907-2ug
Additional Details : Weight : 0.01000kg
Specifications
FTSJ2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSYRCRSA FKLLEVNERH QILRPGLRVL DCGAAPGAWS QVAVQKVNAA GTDPSSPVGF VLGVDLLHIF PLEGATFLCP ADVTDPRTSQ RILEVLPGRR ADVILSDMAP NATGFRDLDH DRLISLCLTL LSVTPDILQP GGTFLCKTWA GSQSRRLQRR LTEEFQNVRI IKPEASRKES SEVYFLATQY HGRKGTVKQ | |
Greater than 95% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FTSJ2 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 30% glycerol. |