missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fusion glycoprotein F0 Protein

A DNA sequence encoding the Homo sapiens (Human) Fusion glycoprotein F0, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9634-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048541

  • 615.00 € / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Fusion glycoprotein F0 Protein
Research Use Only
F
Recombinant Protein
Yeast
NITEEFYQST CSAVSKGYLS ALRTGWYTSV ITIELSNIKE NKCNGTDAKV KLIKQELDKY KNAVTELQLL MQSTPPTNNR ARRELPRFMN YTLNNAKKTN VTLSKKRKRR FLGFLLGVGS AIASGVAVSK VLHLEGEVNK IKSALLSTNK AVVSLSNGVS VLTSKVLDLK NYIDKQLLPI VNKQSCSISN IETVIEFQQK NNRLLEITRE FSVNAGVTTP VSTYMLTNSE LLSLINDMPI TNDQKKLMSN NVQIVRQQSY SIMSIIKEEV LAYVVQLPLY GVIDTPCWKL HTSPLCTTNT KEGSNICLTR TDRGWYCDNA GSVSFFPQAE TCKVQSNRVF CDTMNSLTLP SEINLCNVDI FNPKYDCKIM TSKTDVSSSV ITSLGAIVSC YGKTKCTASN KNRGIIKTFS NGCDYVSNKG MDTVSVGNTL YYVNKQEGKS LYVKGEPIIN FYDPLVFPSD EFDASISQVN EKINQSLAFI RKSDELLHNV NAGKSTTNIM ITT
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers