missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GADD45A / DDIT-1 Protein

A DNA sequence encoding the Homo sapiens (Human) GADD45A / DDIT-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6063-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989709

  • 2285.00 € / 1mg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001186670.1
GADD45A / DDIT-1 Protein
Research Use Only
GADD45A
Recombinant Protein
E. coli
MTLEEFSAGE QKTERMDKVG DALEEVLSKA LSQRTITVGV YEAAKLLNVD PDNVVLCLLA ADEDDDRDVA LQIHFTLIQA FCCENDINIL RVSNPGRLAE LLLLETDAGP AASEGAEQPP DLHCVLVTNP HSSQWKDPAL SQLICFCRES RYMDQWVPVI NLPER
Greater than 90% as determined by SDS-PAGE.
1647
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers