missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GADD45G / CR6 Protein

A DNA sequence encoding the Homo sapiens (Human) GADD45G / CR6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6064-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929719

  • 505.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006696.1
GADD45G / CR6 Protein
Research Use Only
GADD45G
Recombinant Protein
E. coli
MTLEEVRGQD TVPESTARMQ GAGKALHELL LSAQRQGCLT AGVYESAKVL NVDPDNVTFC VLAAGEEDEG DIALQIHFTL IQAFCCENDI DIVRVGDVQR LAAIVGAGEE AGAPGDLHCI LISNPNEDAW KDPALEKLSL FCEESRSVND WVPSITLPE
Greater than 90% as determined by SDS-PAGE.
10912
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers