missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GAGE2A Protein
A cDNA sequence encoding the GAGE2A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11924-20ug
Additional Details : Weight : 0.01000kg
Specifications
GAGE2A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSWRGRS TYRPRPRRYV EPPEMIGPMR PEQFSDEVEP ATPEEGEPAT QRQDPAAAQE GQDEGASAGQ GPKPEAHSQE QGHPQTGCEC EDGPDGQEMD PPNPEEVKTP EEGEKQSQC | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GAGE2A solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol. |