missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GALNT13 Protein

Product Code. 18797379
Change view
Click to view available options
Quantity:
20 μg
Tamaño de la unidad:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
18797379 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18797379 Proveedor R&D Systems™ N.º de proveedor 8906GT020
 Restricted Item
Estimated Shipment: 03-12-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to transfer GalNAc from UDP-GalNAc to peptide EA2 from AnaSpec, Inc. The specific activity is >600 pmol/min/μg, as measured under the described specifications.
TRUSTED_SUSTAINABILITY

Especificaciones

Accession Number Q8IUC8
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 114805
Molecular Weight (g/mol) M.W. (Observed): 60-70 kDa, reducing conditions; M.W. (theoretical): 61 kDa
Quantity 20 μg
Source Human embryonic kidney cell, HEK293-derived human Polypeptide GalNac Transferase 13/GALNT13 protein Ser29-Thr556, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias Acetylgalactosaminyltransferase 13, EC 2.4.1.41, FLJ16031, FLJ41157, GalNAc transferase 13, GalNAc-T13, GalNAc-T13MGC119459, KIAA1918H_NH0187G20.1, MGC119461, Polypeptide GalNAc transferase 13, polypeptide N-acetylgalactosaminyltransferase 13, Pp-GaNTase 13, Protein-UDP acetylgalactosaminyltransferase 13, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 13, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 13 (GalNAc-T13), WUGSC:H_NH0187G20.1
Common Name GALNT13
Gene Symbol GALNT13
Biological Activity Measured by its ability to transfer GalNAc from UDP-GalNAc to peptide EA2 from AnaSpec, Inc. The specific activity is >600 pmol/min/μg, as measured under the described specifications. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Ser29-Thr556, with a C-terminal 6-His tag
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein GALNT13
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.