missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GAPDH Protein

Product Code. 15949759
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15949759 100 μg 100µg
15959759 10 μg 10µg
15999759 50 μg 50µg
15979759 200 μg 200µg
15989759 500 μg 500µg
15969759 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15949759 Supplier enQuireBio™ Supplier No. QP6070ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) GAPDH, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001243728.1
Gene ID (Entrez) 2597
Name GAPDH Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol GAPDH
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence GKVKVGVNGF GRIGRLVTRA AFNSGKVDIV AINDPFIDLN YMVYMFQYDS THGKFHGTVK AENGKLVING NPITIFQERD PSKIKWGDAG AEYVVESTGV FTTMEKAGAH LQGGAKRVII SAPSADAPMF VMGVNHEKYD NSLKIISNAS CTTNCLAPLA KVIHDNFGIV EGLMTTVHAI TATQKTVDGP SGKLWRDGRG ALQNIIPAST GAAKAVGKVI PELNGKLTGM AFRVPTANVS VVDLTCRLEK PAKYDDIKKV VKQASEGPLK GILGYTEHQV VSSDFNSDTH SSTFDAGAGI ALNDHFVKLI SWYDNEFGYS NRVVDLMAHM ASKE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.