missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GDF-9 Protein

Product Code. 18785607
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18785607

Brand: R&D Systems™ 8266G9010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human GDF-9 Protein is derived from HEK293. The Recombinant Human GDF-9 Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature. Upon receipt, store it immediately

TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60383
Formulation Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Molecular Weight (g/mol) 16 kDa (monomer)
Name GDF-9
Quantity 10 μg
Source Human embryonic kidney cell,HEK293-derived human GDF-9 protein Gly320-Arg454 (Gly391Arg)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.