missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GGPS1 / GGPPS1 Protein

A DNA sequence encoding the Homo sapiens (Human) GGPS1 / GGPPS1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6082-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929839

  • 458.00 € / 100µg
Estimated Shipment: 23-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001032354.1
GGPS1 / GGPPS1 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
9453
Greater than 80.8% as determined by SDS-PAGE.
Research Use Only
GGPS1
Recombinant Protein
E. coli
MEKTQETVQR ILLEPYKYLL QLPGKQVRTK LSQAFNHWLK VPEDKLQIII EVTEMLHNAS LLIDDIEDNS KLRRGFPVAH SIYGIPSVIN SANYVYFLGL EKVLTLDHPD AVKLFTRQLL ELHQGQGLDI YWRDNYTCPT EEEYKAMVLQ KTGGLFGLAV GLMQLFSDYK EDLKPLLNTL GLFFQIRDDY ANLHSKEYSE NKSFCEDLTE GKFSFPTIHA IWSRPESTQV QNILRQRTEN IDIKKYCVHY LEDVGSFEYT RNTLKELEAK AYKQIDARGG NPELVALVKH LSKMFKEENE
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers