missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GH1 / Growth hormone 1 Protein
Recombinant Protein
Brand: enQuireBio™ QP5354-5mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000506.2 | |
GH1 / Growth hormone 1 Protein | |
5 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human GH1 / Growth hormone 1 induces PDF 9D11 cell proliferation (typical ED50 is 1-15 ng/ml). | |
Human | |
Untagged | |
20 mM sodium bicarbonate, pH 8.0 |
2688 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
GH1 | |
Recombinant Protein | |
E. coli | |
MFPTIPLSRL FDNAMLRAHR LHQLAFDTYQ EFEEAYIPKE QKYSFLQNPQ TSLCFSESIP TPSNREETQQ KSNLELLRIS LLLIQSWLEP VQFLRSVFAN SLVYGASDSN VYDLLKDLEE GIQTLMGRLE DGSPRTGQIF KQTYSKFDTN SHNDDALLKN YGLLYCFRKD MDKVETFLRI VQCRSVEGSC GF |