missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GITR Ligand/TNFSF18 (CHO-expressed)

Product Code. 18320538
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18320538

Brand: R&D Systems™ 6987GL025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to induce IL-8 secretion in HT1080 human fibrosarcoma cells transfected with GITR. Wyzgol, A. et al. (2009) J. Immunol. 183:1851. The ED50 for this effect is 4-20 ng/mL. The activity can be enhanced approximately 10-fold in the presence of Mouse Anti-Hemagglutinin/HA Peptide Monoclonal Antibody (Catalog # MAB060 ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_005083
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8995
Molecular Weight (g/mol) M.W. (Observed): 21-30 kDa, reducing conditions; M.W. (theoretical): 19.6 kDa (monomer)
Name GITR Ligand/TNFSF18
Quantity 25 μg
Source Chinese Hamster Ovary cell line, CHO-derived human GITR Ligand/TNFSF18 protein Hemagglutinin Tag (YPYDVPDYA) GCN4-IZ GGGSGGGSGGGS Human GITR Ligand (Glu74-Ser199) (N-terminus) Accession # NP_005083 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Non-covalently-linked trimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Gene Symbol TNFSF18
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein GITR Ligand/TNFSF18
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.