missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GKN1 Protein
A cDNA sequence encoding the GKN1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11977-100ug
Additional Details : Weight : 0.01000kg
Specifications
56287 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSH MLAYSS VHCFREDKMK FTIVFAGLLG VFLAPALANY NINVNDDNNN AGSGQQSVSV NNEHNVANVD NNNGWDSWNS IWDYGNGFAA TRLFQKKTCI VHKMNKEVMP SIQSLDALVK EKKLQGKGPG GPPPKGLMYS VNPNKVDDLS KFGKNIANMC RGIPTYMAEE MQEASLFFYS GTCYTTSVLW IVDISFCGDT VEN | |
Greater than 85% as determined by SDS-PAGE. |
GKN1 Protein | |
Research Use Only | |
GKN1 | |
Human | |
His | |
The GKN1 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |