missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GKN1 Protein

A cDNA sequence encoding the GKN1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11977-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928988

  • 131.00 € / 2µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

56287
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSH MLAYSS VHCFREDKMK FTIVFAGLLG VFLAPALANY NINVNDDNNN AGSGQQSVSV NNEHNVANVD NNNGWDSWNS IWDYGNGFAA TRLFQKKTCI VHKMNKEVMP SIQSLDALVK EKKLQGKGPG GPPPKGLMYS VNPNKVDDLS KFGKNIANMC RGIPTYMAEE MQEASLFFYS GTCYTTSVLW IVDISFCGDT VEN
Greater than 85% as determined by SDS-PAGE.
GKN1 Protein
Research Use Only
GKN1
Human
His
The GKN1 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers