missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLOD4 Protein
A cDNA sequence encoding the GLOD4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11988-1mg
Additional Details : Weight : 0.01000kg
Specifications
GLOD4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAARRALHFV FKVGNRFQTA RFYRDVLGMK VLRHEEFEEG CKAACNGPYD GKWSKTMVGF GPEDDHFVAE LTYNYGVGDY KLGNDFMGIT LASSQAVSNA RKLEWPLTEV AEGVFETEAP GGYKFYLQNR SLPQSDPVLK VTLAVSDLQK SLNYWCNLLG MKIYEKDEEK QRALLGYADN QCKLELQGVK GGVDHAAAFG RIAFSCPQKE LPDLEDLMKR ENQKILTPLV SLDTPGKATV QVVILADPDG HEICFVGDEA FRELSKMDPE GSKLLDDAMA ADKSDEWFAK HNKPKASG | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GLOD4 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 20% glycerol. |