missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glutathione peroxidase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Glutathione peroxidase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6115-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050910

  • 505.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000572.2
Glutathione peroxidase 1 Protein
Research Use Only
GPX1
Recombinant Protein
E. coli
MCAARLAAAA AAAQSVYAFS ARPLAGGEPV SLGSLRGKVL LIENVASLUG TTVRDYTQMN ELQRRLGPRG LVVLGFPCNQ FGHQENAKNE EILNSLKYVR PGGGFEPNFM LFEKCEVNGA GAHPLFAFLR EALPAPSDDA TALMTDPKLI TWSPVCRNDV AWNFEKFLVG PDGVPLRRYS RRFQTIDIEP DIEALLSQGP SCA
Greater than 90% as determined by SDS-PAGE.
2876
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers