missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutathione peroxidase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutathione peroxidase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6115-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000572.2 | |
Glutathione peroxidase 1 Protein | |
Research Use Only | |
GPX1 | |
Recombinant Protein | |
E. coli | |
MCAARLAAAA AAAQSVYAFS ARPLAGGEPV SLGSLRGKVL LIENVASLUG TTVRDYTQMN ELQRRLGPRG LVVLGFPCNQ FGHQENAKNE EILNSLKYVR PGGGFEPNFM LFEKCEVNGA GAHPLFAFLR EALPAPSDDA TALMTDPKLI TWSPVCRNDV AWNFEKFLVG PDGVPLRRYS RRFQTIDIEP DIEALLSQGP SCA | |
Greater than 90% as determined by SDS-PAGE. |
2876 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |