missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutathione S-transferase A2 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutathione S-transferase A2, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_000837.3 |
---|---|
Gene ID (Entrez) | 2939 |
Name | Glutathione S-transferase A2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16010980
|
enQuireBio™
QP6123-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16030980
|
enQuireBio™
QP6123-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020980
|
enQuireBio™
QP6123-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040980
|
enQuireBio™
QP6123-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050980
|
enQuireBio™
QP6123-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060980
|
enQuireBio™
QP6123-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000837.3 | |
Glutathione S-transferase A2 Protein | |
Not determined. | |
Full Length of BC002895 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
2939 | |
Research Use Only | |
GSTA2 | |
Recombinant Protein | |
E. coli | |
MAEKPKLHYS NIRGRMESIR WLLAAAGVEF EEKFIKSAED LDKLRNDGYL MFQQVPMVEI DGMKLVQTRA ILNYIASKYN LYGKDIKEKA LIDMYIEGIA DLGEMILLLP FTQPEEQDAK LALIQEKTKN RYFPAFEKVL KSHGQDYLVG NKLSRADIHL VELLYYVEEL DSSLISSFPL LKALKTRISN LPTVKKFLQP GSPRKPPMDE KSLEESRKIF RF | |
Greater than 80% as determined by SDS-PAGE. |