missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glycine N-acyltransferase-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Glycine N-acyltransferase-like protein 1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001207423.1 |
---|---|
Gene ID (Entrez) | 92292 |
Name | Glycine N-acyltransferase-like protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051441
|
enQuireBio™
QP7893-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071441
|
enQuireBio™
QP7893-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061441
|
enQuireBio™
QP7893-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081441
|
enQuireBio™
QP7893-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091441
|
enQuireBio™
QP7893-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001451
|
enQuireBio™
QP7893-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001207423.1 | |
Glycine N-acyltransferase-like protein 1 | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
92292 | |
Research Use Only | |
GLYATL1 | |
Recombinant Protein | |
E. coli | |
MILLNNSHKL LALYKSLARS IPESLKVYGS VYHINHGNPF NMEVLVDSWP EYQMVIIRPQ KQEMTDDMDS YTNVYRMFSK EPQKSEEVLK NCEIVNWKQR LQIQGLQESL GEGIRVATFS KSVKVEHSRA LLLVTEDILK LNASSKSKLG SWAETGHPDD EFESETPNFK YAQLDVSYSG LVNDNWKRGK NERSLHYIKR CIEDLPAACM LGPEGVPVSW VTMDPSCEVG MAYSMEKYRR TGNMARVMVR YMKYLRQKNI PFYISVLEEN EDSRRFVGQF GFFEASCEWH QWTCYPQNLV PF | |
Greater than 80% as determined by SDS-PAGE. |