missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glycogen phosphorylase, brain form Protein
A DNA sequence encoding the Homo sapiens (Human) Glycogen phosphorylase, brain form, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_002853.2 |
---|---|
Gene ID (Entrez) | 5834 |
Name | Glycogen phosphorylase, brain form Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16045931
|
enQuireBio™
QP8681-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065931
|
enQuireBio™
QP8681-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055931
|
enQuireBio™
QP8681-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075931
|
enQuireBio™
QP8681-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085931
|
enQuireBio™
QP8681-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095931
|
enQuireBio™
QP8681-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002853.2 | |
Glycogen phosphorylase, brain form Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5834 | |
Research Use Only | |
PYGB | |
Recombinant Protein | |
E. coli | |
TDSEKRKQIS VRGLAGLGDV AEVRKSFNRH LHFTLVKDRN VATPRDYFFA LAHTVRDHLV GRWIRTQQHY YERDPKRIYY LSLEFYMGRT LQNTMVNLGL QNACDEAIYQ LGLDLEELEE IEEDAGLGNG GLGRLAACFL DSMATLGLAA YGYGIRYEFG IFNQKIVNGW QVEEADDWLR YGNPWEKARP EYMLPVHFYG RVEHTPDGVK WLDTQVVLAM PYDTPVPGYK NNTVNTMRLW SAKAPNDFKL QDFNVGDYIE AVLDRNLAEN ISRVLYPNDN FFEGKELRLK QEYFVVAATL QDIIRRFKSS KFGCRDPVRT CFETFPDKVA IQLNDTHPAL SIPELMRILV DVEKVDWDKA WEITKKTCAY TNHTVLPEAL ERWPVSMFEK LLPRHLEIIY AINQRHLDHV AALFPGDVDR LRRMSVIEEG DCKRINMAHL CVIGSHAVNG VARIHSEIVK QSVFKDFYEL EPEKFQNKTN GITPRRWLLL CNPGLADTIV EKIGEEFLTD LSQLKKLLPL VSDEVFIRDV AKVKQENKLK FSAFLEKEYK VKINPSSMFD VHVKRIHEYK RQLLNCLHVV TLYNRIKRDP AKAFVPRTVM IGGKAAPGYH MAKLIIKLVT SIGDVVNHDP VVGDRLKVIF LENYRVSLAE KVIPAADLSQ QISTAGTEAS GTGNMKFMLN GALTIGTMDG ANVEMAEEAG AENLFIFGLR VEDVEALDRK GYNAREYYDH LPELKQAVDQ ISSGFFSPKE PDCFKDIVNM LMHHDRFKVF ADYEAYMQCQ AQVDQLYRNP KEWTKKVIRN IACSGKFSSD RTITEYAREI WGVEPSDLQI PPPNIPRD | |
Greater than 90% as determined by SDS-PAGE. |