missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GMDS Protein

Product Code. p-7149854
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15939058

Brand: enQuireBio™ QP120015ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GMDS was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 2762
Name GMDS Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol GMDS
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MAHAPARCPS ARGSGDGEMG KPRNVALITG ITGQDGSYLA EFLLEKGYEV HGIVRRSSSF NTGRIEHLYK NPQAHIEGNM KLHYGDLTDS TCLVKIINEV KPTEIYNLGA QSHVKISFDL AEYTADVDGV GTLRLLDAVK TCGLINSVKF YQASTSELYG KVQEIPQKET TPFYPRSPYG AAKLYAYWIV VNFREAYNLF AVNGILFNHE SPRRGANFVT RKISRSVAKI YLGQLECFSL GNLDAKRDWG HAKDYVEAMW LMLQNDEPED FVIATGEVHS VREFVEKSFL HIGKTIVWEG KNENEVGRCK ETGKVHVTVD LKYYRPTEVD FLQGDCTKAK QKLNWKPRVA FDELVREMVH ADVELMRTNP NA
Buffer The GMDS solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl, 0.1mM PMSF and 30% glycerol.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.