missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GMP reductase 2 Protein

Product Code. 16022811
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16022811 1 mg 1mg
16012811 10 μg 10µg
16052811 50 μg 50µg
16002811 100 μg 100µg
16032811 200 μg 200µg
16042811 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16022811 Supplier enQuireBio™ Supplier No. QP8126ec1mg
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) GMP reductase 2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001002000.1
Gene ID (Entrez) 51292
Name GMP reductase 2 Protein
Quantity 1 mg
Regulatory Status RUO
Endotoxin Concentration Not determined.
Gene Symbol GMPR2
Protein Length Full Length of Isoform 2
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MTSCLPALRF IATPRLSAMP HIDNDVKLDF KDVLLRPKRS TLKSRSEVDL TRSFSFRNSK QTYSGVPIIA ANMDTVGTFE MAKVLCKFSL FTAVHKHYSL VQWQEFAGQN PDCLEHLAAS SGTGSSDFEQ LEQILEAIPQ VKYICLDVAN GYSEHFVEFV KDVRKRFPQH TIMAGNVVTG EMVEELILSG ADIIKVGIGP GSVCTTRKKT GVGYPQLSAV MECADAAHGL KGHIISDGGC SCPGDVAKAF GAGADFVMLG GMLAGHSESG GELIERDGKK YKLFYGMSSE MAMKKYAGGV AEYRASEGKT VEVPFKGDVE HTIRDILGGI RSTCTYVGAA KLKELSRRTT FIRVTQQVNP IFSEAC
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.