missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GNG13 Protein

Product Code. p-7149872
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
This item is not returnable. View return policy

Product Code. 15969108

Brand: enQuireBio™ QP120182ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GNG13 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 51764
Name GNG13 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol GNG13
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMEEWDVP QMKKEVESLK YQLAFQREMA SKTIPELLKW IEDGIPKDPF LNPDLMKNNP WVEKGKC
Buffer GNG13 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol, 2mM DTT and 0.1mM PMSF.
Purity or Quality Grade Greater than 80.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.