missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GNPNAT1 Protein

A cDNA sequence encoding the GNPNAT1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12023-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15999118

  • 131.00 € / 2µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GNPNAT1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMKPDETP MFDPSLLKEV DWSQNTATFS PAISPTHPGE GLVLRPLCTA DLNRGFFKVL GQLTETGVVS PEQFMKSFEH MKKSGDYYVT VVEDVTLGQI VATATLIIEH KFIHSCAKRG RVEDVVVSDE CRGKQLGKLL LSTLTLLSKK LNCYKITLEC LPQNVGFYKK FGYTVSEENY MCRRFLK
Greater than 90% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GNPNAT1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 30% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers