missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GPD1L Protein
A cDNA sequence encoding the GPD1L was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12036-20ug
Additional Details : Weight : 0.01000kg
Specifications
23171 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAAPLKVCI VGSGNWGSAV AKIIGNNVKK LQKFASTVKM WVFEETVNGR KLTDIINNDH ENVKYLPGHK LPENVVAMSN LSEAVQDADL LVFVIPHQFI HRICDEITGR VPKKALGITL IKGIDEGPEG LKLISDIIRE KMGIDISVLM GANIANEVAA EKFCETTIGS KVMENGLLFK ELLQTPNFRI TVVDDADTVE LCGALKNIVA VGAGFCDGLR CGDNTKAAVI RLGLMEMIAF ARIFCKGQVS TATFLESCGV ADLITTCYGG RNRRVAEAFA RTGKTIEELE KEMLNGQKLQ GPQTSAEVYR ILKQKGLLDK FPLFTAVYQI CYESRPVQEM LSCLQSHPEH T | |
Greater than 95.0% as determined by SDS-PAGE. |
GPD1L Protein | |
Research Use Only | |
GPD1L | |
Human | |
His | |
GPD1L protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT. |