missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GPNMB Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | GPNMB Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | Human (HEK293) |
| Protein Tag | His |
| Sequence | AKRFHDVLGN ERPSAYMREH NQLNGWSSDE NDWNEKLYPV WKRGDMRWKN SWKGGRVQAV LTSDSPALVG SNITFAVNLI FPRCQKEDAN GNIVYEKNCR NEAGLSADPY VYNWTAWSED SDGENGTGQS HHNVFPDGKP FPHHPGWRRW NFIYVFHTLG QYFQKLGRCS VRVSVNTANV TLGPQLMEVT VYRRHGRAYV PIAQVKDVYV VTDQIPVFVT MFQKNDRNSS DETFLKDLPI MFDVLIHDPS HFLNYSTINY KWSFGDNTGL FVSTNHTVNH TYVLNGTFSL NLTVKAAAPG PCPPPPPPPR PSKPTPSLAT TLKSYDSNTP GPAGDNPLEL SRIPDENCQI NRYGHFQATI TIVEGILEVN IIQMTDVLMP VPWPESSLID FVVTCQGSIP TEVCTIISDP TCEITQNTVC SPVDVDEMCL LTVRRTFNGS GTYCVNLTLG DDTSLALTST LISVP KLHHH HHHHHHH |
| Buffer | Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction