missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GPNMB Protein

Product Code. p-7149896
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15979178

Brand: enQuireBio™ QP12041HISINSECT2ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GPNMB was constructed and used to recombinantly synthesize the protein.

Specifications

Name GPNMB Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence AKRFHDVLGN ERPSAYMREH NQLNGWSSDE NDWNEKLYPV WKRGDMRWKN SWKGGRVQAV LTSDSPALVG SNITFAVNLI FPRCQKEDAN GNIVYEKNCR NEAGLSADPY VYNWTAWSED SDGENGTGQS HHNVFPDGKP FPHHPGWRRW NFIYVFHTLG QYFQKLGRCS VRVSVNTANV TLGPQLMEVT VYRRHGRAYV PIAQVKDVYV VTDQIPVFVT MFQKNDRNSS DETFLKDLPI MFDVLIHDPS HFLNYSTINY KWSFGDNTGL FVSTNHTVNH TYVLNGTFSL NLTVKAAAPG PCPPPPPPPR PSKPTPSLAT TLKSYDSNTP GPAGDNPLEL SRIPDENCQI NRYGHFQATI TIVEGILEVN IIQMTDVLMP VPWPESSLID FVVTCQGSIP TEVCTIISDP TCEITQNTVC SPVDVDEMCL LTVRRTFNGS GTYCVNLTLG DDTSLALTST LISVP KLHHH HHHHHHH
Buffer Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.