missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Growth Hormone (GH) Protein

Product Code. 18176500
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
Unit Size:
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation Unit Size
18176500 Lyophilized from a 0.2μm filtered solution in Tris and NaCl. 25µg
18118040 Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein. 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18176500 Supplier R&D Systems™ Supplier No. 1067GH025/CF
  • ONLINE EXCLUSIVE PRICE
    261.91 €
    233.10 € / 25µg
    Save 28.81 €
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Growth Hormone (GH) Protein is derived from E. coli. The Recombinant Human Growth Hormone (GH) Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 2688
Molecular Weight (g/mol) 22 kDa
Name Growth Hormone
Quantity 25 μg
Source E. coli-derived human Growth Hormone protein Phe27-Phe217,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.