missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GSTM3 Protein

A cDNA sequence encoding the GSTM3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12058-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959228

  • 203.00 € / 20µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GSTM3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMSCESS MVLGYWDIRG LAHAIRLLLE FTDTSYEEKR YTCGEAPDYD RSQWLDVKFK LDLDFPNLPY LLDGKNKITQ SNAILRYIAR KHNMCGETEE EKIRVDIIEN QVMDFRTQLI RLCYSSDHEK LKPQYLEELP GQLKQFSMFL GKFSWFAGEK LTFVDFLTYD ILDQNRIFDP KCLDEFPNLK AFMCRFEALE KIAAYLQSDQ FCKMPINNKM AQWGNKPVC
Greater than 95% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GSTM3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl,1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers