missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GSTM5 Protein

A cDNA sequence encoding the GSTM5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10660-1mg


Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954558

  • 7540.00 € / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GSTM5 Protein
Research Use Only
Specific activity is > 90,000 pmol/min/ug, and is defined as the amount of enzyme that conjugate 1.0 u mole of 1-chloro-2,4- dinitrobenzene (CDNB) with reduced glutathione per minute at pH 6.5 at 25C.
Human
His
The GSTM5 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMPMTLG YWDIRGLAHA IRLLLEYTDS SYVEKKYTLG DAPDYDRSQW LNEKFKLGLD FPNLPYLIDG AHKITQSNAI LRYIARKHNL CGETEEEKIR VDILENQVMD NHMELVRLCY DPDFEKLKPK YLEELPEKLK LYSEFLGKRP WFAGDKITFV DFLAYDVLDM KRIFEPKCLD AFLNLKDFIS RFEGLKKISA YMKSSQFLRG LLFGKSATWN SK
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers