missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GSTO1 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | GSTO1 Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MSGESARSLG KGSAPPGPVP EGSIRIYSMR FCPFAERTRL VLKAKGIRHE VININLKNKP EWFFKKNPFG LVPVLENSQG QLIYESAITC EYLDEAYPGK KLLPDDPYEK ACQKMILELF SKVPSLVGSF IRSQNKEDYA GLKEEFRKEF TKLEEVLTNK KTTFFGGNSI SMIDYLIWPW FERLEAMKLN ECVDHTPKLK LWMAAMKEDP TVSALLTSEK DWQGFLELYL QNSPEACDYG L |
| Buffer | GSTO1 protein is supplied in 20 mM Tris pH 8.0 and 50% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction