missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GSTO2 Protein
A cDNA sequence encoding the GSTO2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12062-10ug
Additional Details : Weight : 0.01000kg
Specifications
GSTO2 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GSTO2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 40% glycerol. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSGDATR TLGKGSQPPG PVPEGLIRIY SMRFCPYSHR TRLVLKAKDI RHEVVNINLR NKPEWYYTKH PFGHIPVLET SQCQLIYESV IACEYLDDAY PGRKLFPYDP YERARQKMLL ELFCKVPHLT KECLVALRCG RECTNLKAAL RQEFSNLEEI LEYQNTTFFG GTCISMIDYL LWPWFERLDV YGILDCVSHT PALRLWISAM KWDPTVCALL MDKSIFQGFL NLYFQNNPNA FDFGLC |