missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GTF2A2 Protein

A DNA sequence encoding the Homo sapiens (Human) GTF2A2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6129-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051020

  • 2285.00 € / 1mg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001307858.1
GTF2A2 Protein
Research Use Only
GTF2A2
Recombinant Protein
E. coli
MAYQLYRNTT LGNSLQESLD ELIQSQQITP QLALQVLLQF DKAINAALAQ RVRNRVNFRG SLNTYRFCDN VWTFVLNDVE FREVTELIKV DKVKIVACDG KNTGSNTTE
Greater than 80% as determined by SDS-PAGE.
2958
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers