missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Guanylate cyclase activator 2B Protein
A DNA sequence encoding the Homo sapiens (Human) Guanylate cyclase activator 2B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7537-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_009033.1 | |
Guanylate cyclase activator 2B Protein | |
Research Use Only | |
Guca2b | |
Recombinant Protein | |
E. coli | |
VYIQYQGFRV QLESMKKLSD LEAQWAPSPR LQAQSLLPAV CHHPALPQDL QPVCASQEAS SIFKTLRTIA NDDCELCVNV ACTGCL | |
Greater than 90% as determined by SDS-PAGE. |
2981 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |