missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Guanylate kinase Protein
A DNA sequence encoding the Homo sapiens (Human) Guanylate kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7566-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000849.1 | |
Guanylate kinase Protein | |
Research Use Only | |
GUK1 | |
Recombinant Protein | |
E. coli | |
SGPRPVVLSG PSGAGKSTLL KRLLQEHSGI FGFSVSHTTR NPRPGEENGK DYYFVTREVM QRDIAAGDFI EHAEFSGNLY GTSKVAVQAV QAMNRICVLD VDLQGVRNIK ATDLRPIYIS VQPPSLHVLE QRLRQRNTET EESLVKRLAA AQADMESSKE PGLFDVVIIN DSLDQAYAEL KEALSEEIKK AQRTGA | |
Greater than 90% as determined by SDS-PAGE. |
2987 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |