missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Guanylate kinase Protein

A DNA sequence encoding the Homo sapiens (Human) Guanylate kinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7566-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049440

  • 2285.00 € / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000849.1
Guanylate kinase Protein
Research Use Only
GUK1
Recombinant Protein
E. coli
SGPRPVVLSG PSGAGKSTLL KRLLQEHSGI FGFSVSHTTR NPRPGEENGK DYYFVTREVM QRDIAAGDFI EHAEFSGNLY GTSKVAVQAV QAMNRICVLD VDLQGVRNIK ATDLRPIYIS VQPPSLHVLE QRLRQRNTET EESLVKRLAA AQADMESSKE PGLFDVVIIN DSLDQAYAEL KEALSEEIKK AQRTGA
Greater than 90% as determined by SDS-PAGE.
2987
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers